Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR89A
Entrez Gene ID
483157
Gene Name
G protein-coupled receptor 89A
Gene Aliases
GPR89B
Chromosome Location
Chr.17: 58436920 - 58498082 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630713.3 XP_005630770.1 6 - 7 695 70
XM_022404930.1 XP_022260638.1 6 - 7 697 70
XM_022404931.1 XP_022260639.1 6 - 7 695 70
XM_022404932.1 XP_022260640.1 5 - 6 591 70
XM_844483.5 XP_849576.1 6 - 7 695 70

View Full Product Details