Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOSTDC1
Entrez Gene ID
100856691
Gene Name
sclerostin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 31022599 - 31027155 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639481.4 XP_003639529.1 2 - 2 853 153

View Full Product Details