Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
RTKN2
Entrez Gene ID
479218
Gene Name
rhotekin 2
Gene Aliases
PLEKHK1
Chromosome Location
Chr.4: 14165569 - 14245088 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618914.2 XP_005618971.1 9 - 9 946 143
XM_005618915.3 XP_005618972.1 9 - 9 947 143
XM_005618917.3 XP_005618974.1 9 - 9 947 143
XM_005618918.3 XP_005618975.1 9 - 9 946 143
XM_005618919.3 XP_005618976.1 9 - 9 946 143
XM_014112826.2 XP_013968301.1 9 - 9 1124 143
XM_022417459.1 XP_022273167.1 9 - 9 851 143
XM_022417460.1 XP_022273168.1 9 - 9 1045 143
XM_536360.5 XP_536360.3 9 - 9 946 143

View Full Product Details