Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CD6
Entrez Gene ID
483804
Gene Name
CD6 molecule
Gene Aliases
-
Chromosome Location
Chr.18: 55296116 - 55336014 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631691.3 XP_005631748.1 12 - 12 2215 104
XM_014121151.2 XP_013976626.1 12 - 12 2586 104
XM_014121152.2 XP_013976627.1 12 - 12 2582 101
XM_014121153.2 XP_013976628.1 11 - 11 2502 104
XM_014121154.2 XP_013976629.1 10 - 10 2391 104
XM_014121155.2 XP_013976630.1 11 - 11 2291 104
XM_014121156.2 XP_013976631.1 11 - 11 2496 101

View Full Product Details