Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRIF1
Entrez Gene ID
479906
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
-
Chromosome Location
Chr.6: 41023383 - 41041776 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621791.3 XP_005621848.1 4 - 4 2616 99
XM_022420340.1 XP_022276048.1 4 - 4 2436 99
XM_860702.5 XP_865795.1 3 - 3 933 99

View Full Product Details