Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
479247
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr.4: 23965952 - 23974053 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536389.6 XP_536389.3 3 - 4 1290 106

View Full Product Details