Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A2
Entrez Gene ID
491964
Gene Name
solute carrier family 16 member 2
Gene Aliases
-
Chromosome Location
Chr.X: 57845865 - 57859827 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022415864.1 XP_022271572.1 2 - 3 624 113

View Full Product Details