Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP4B1
Entrez Gene ID
483125
Gene Name
adaptor related protein complex 4 subunit beta 1
Gene Aliases
-
Chromosome Location
Chr.17: 51701663 - 51732022 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630636.3 XP_005630693.1 10 - 11 2179 155
XM_005630638.3 XP_005630695.1 9 - 10 1633 155
XM_014120611.2 XP_013976086.1 9 - 10 1664 155

View Full Product Details