Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
TNFAIP6
Entrez Gene ID
476147
Gene Name
TNF alpha induced protein 6
Gene Aliases
-
Chromosome Location
Chr.19: 52607598 - 52623799 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533354.6 XP_533354.3 3 - 3 484 164

View Full Product Details