Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHOD
Entrez Gene ID
611862
Gene Name
ras homolog family member D
Gene Aliases
-
Chromosome Location
Chr.18: 50320428 - 50330108 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022405606.1 XP_022261314.1 3 - 4 357 161
XM_849583.6 XP_854676.3 3 - 4 357 161

View Full Product Details