Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YJU2
Entrez Gene ID
611929
Gene Name
YJU2 splicing factor homolog
Gene Aliases
CCDC94
Chromosome Location
Chr.20: 55367512 - 55380755 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005633058.3 XP_005633115.1 4 - 5 539 101
XM_022407213.1 XP_022262921.1 4 - 5 539 101

View Full Product Details