Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
610152
Gene Name
vesicle amine transport 1 like
Gene Aliases
-
Chromosome Location
Chr.5: 73330322 - 73476928 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847568.5 XP_852661.2 8 - 9 1287 141

View Full Product Details