Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPSM3
Entrez Gene ID
607687
Gene Name
G protein signaling modulator 3
Gene Aliases
-
Chromosome Location
Chr.12: 1583438 - 1585677 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014118355.2 XP_013973830.1 2 - 3 643 137
XM_844458.5 XP_849551.1 2 - 3 634 137

View Full Product Details