Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR52P1
Entrez Gene ID
485292
Gene Name
olfactory receptor family 52 subfamily P member 1
Gene Aliases
cOR52P1P, cOR52P3
Chromosome Location
Chr.21: 28815755 - 28816735 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017521.1 NP_001017521.1 1 - 1 919 81

View Full Product Details