Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam81a
Entrez Gene ID
100774523
Gene Name
family with sequence similarity 81, member A
Gene Aliases
I79_000181
Chromosome Location
Chr.Un|NW_003613582: 6189626 - 6223968 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495044.2 XP_003495092.1 1 - 2 287 63

View Full Product Details