Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpy19l2
Entrez Gene ID
100752353
Gene Name
dpy-19-like 2 (C. elegans)
Gene Aliases
I79_018711
Chromosome Location
Chr.Un|NW_003614877: 58863 - 199670 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510425.1 XP_003510473.1 23 - 24 2258 65

View Full Product Details