Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sostdc1
Entrez Gene ID
100758491
Gene Name
sclerostin domain containing 1
Gene Aliases
I79_015619
Chromosome Location
Chr.Un|NW_003614441: 485381 - 488518 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507863.2 XP_003507911.1 1 - 2 205 83

View Full Product Details