Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hfe
Entrez Gene ID
100762132
Gene Name
hemochromatosis
Gene Aliases
I79_008706
Chromosome Location
Chr.Un|NW_003613881: 337531 - 352545 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642819.1 XP_007641009.1 4 - 5 1075 74

View Full Product Details