Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1d
Entrez Gene ID
100761266
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1D
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613806: 1339821 - 1374538 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641482.1 XP_007639672.1 2 - 3 336 82

View Full Product Details