Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Znf76
Entrez Gene ID
100755287
Gene Name
zinc finger protein 76
Gene Aliases
Zfp523
Chromosome Location
Chr.Un|NW_003614227: 99783 - 114987 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506290.2 XP_003506338.1 8 - 9 1036 96
XM_007646796.1 XP_007644986.1 8 - 9 1037 96
GenBank mRNA JI877658.1 - 8 - 9 961 96
JP051495.1 - 8 - 9 961 96

View Full Product Details