Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tyw1
Entrez Gene ID
100751910
Gene Name
tRNA-yW synthesizing protein 1 homolog (S. cerevisiae)
Gene Aliases
I79_023155
Chromosome Location
Chr.Un|NW_003616310: 453 - 61723 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654239.1 XP_007652429.1 3 - 4 328 63

View Full Product Details