Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Podnl1
Entrez Gene ID
100759324
Gene Name
podocan-like 1
Gene Aliases
I79_008374
Chromosome Location
Chr.Un|NW_003613865: 1154092 - 1160349 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501905.2 XP_003501953.1 8 - 9 1524 66

View Full Product Details