Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Efemp1
Entrez Gene ID
100750809
Gene Name
EGF containing fibulin-like extracellular matrix protein 1
Gene Aliases
I79_020269
Chromosome Location
Chr.Un|NW_003615198: 267946 - 323353 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511681.2 XP_003511729.2 - 1023 64
GenBank mRNA JP060013.1 - - 974 64

View Full Product Details