Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k8
Entrez Gene ID
100759538
Gene Name
mitogen-activated protein kinase kinase kinase 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614468: 211384 - 232405 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508000.2 XP_003508048.1 7 - 8 1332 60

View Full Product Details