Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankk1
Entrez Gene ID
100751636
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613694: 2457632 - 2468720 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498541.1 XP_003498589.1 3 - 4 664 84

View Full Product Details