Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp20
Entrez Gene ID
100755720
Gene Name
matrix metallopeptidase 20
Gene Aliases
I79_001479
Chromosome Location
Chr.Un|NW_003613612: 888990 - 921682 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496135.2 XP_003496183.1 7 - 8 1092 62

View Full Product Details