Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmsb15a
Entrez Gene ID
100767589
Gene Name
thymosin beta 15a
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614444: 632322 - 633208 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507888.1 XP_003507936.1 1 - 2 105 115

View Full Product Details