Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh2d6
Entrez Gene ID
100752626
Gene Name
SH2 domain containing 6
Gene Aliases
I79_012881
Chromosome Location
Chr.Un|NW_003614172: 292651 - 299501 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505639.2 XP_003505687.1 10 - 11 751 56

View Full Product Details