Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Surf6
Entrez Gene ID
100771435
Gene Name
surfeit 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615220: 146002 - 151773 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511729.2 XP_003511777.1 4 - 5 661 58
GenBank mRNA JI883990.1 - 4 - 5 648 58
JP055378.1 - 4 - 5 648 58

View Full Product Details