Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Maneal
Entrez Gene ID
100754796
Gene Name
mannosidase, endo-alpha-like
Gene Aliases
I79_008780
Chromosome Location
Chr.Un|NW_003613886: 778665 - 782543 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642859.1 XP_007641049.1 1 - 2 74 70

View Full Product Details