Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poc5
Entrez Gene ID
100772693
Gene Name
POC5 centriolar protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615832: 79662 - 111509 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653578.1 XP_007651768.1 10 - 11 1716 58
XM_007653579.1 XP_007651769.1 10 - 11 1720 58
XM_007653580.1 XP_007651770.1 10 - 11 1605 58
GenBank mRNA JI874376.1 - 4 - 5 693 58
JP066095.1 - 6 - 7 966 58

View Full Product Details