Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap4b1
Entrez Gene ID
100759820
Gene Name
adaptor-related protein complex 4, beta 1 subunit
Gene Aliases
I79_013084
Chromosome Location
Chr.Un|NW_003614192: 789125 - 802217 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505815.2 XP_003505863.1 5 - 6 907 58
XM_007646301.1 XP_007644491.1 5 - 6 767 58

View Full Product Details