Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Susd4
Entrez Gene ID
100771822
Gene Name
sushi domain containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613581: 1097630 - 1155020 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643918.1 XP_007642108.1 1 - 2 33 60

View Full Product Details