Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h3
Entrez Gene ID
100756959
Gene Name
zinc finger CCCH-type containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614880: 67155 - 164615 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510436.2 XP_003510484.2 10 - 11 2632 66
XM_007651069.1 XP_007649259.1 10 - 11 2656 66
XM_007651070.1 XP_007649260.1 9 - 10 2431 66
XM_007651071.1 XP_007649261.1 9 - 10 2406 66
XM_007651072.1 XP_007649262.1 9 - 10 2345 66
XM_007651073.1 XP_007649263.1 9 - 10 2341 66
XM_007651074.1 XP_007649264.1 10 - 11 2656 66
GenBank mRNA JI881380.1 - 5 - 6 607 66
JP046687.1 - 5 - 6 612 66

View Full Product Details