Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp11
Entrez Gene ID
100770939
Gene Name
dual specificity phosphatase 11 (RNA/RNP complex 1-interacting)
Gene Aliases
I79_018276
Chromosome Location
Chr.Un|NW_003614815: 38739 - 54507 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650682.1 XP_007648872.1 7 - 8 699 66
XM_007650683.1 XP_007648873.1 7 - 8 700 66
XM_007650685.1 XP_007648875.1 7 - 8 765 66

View Full Product Details