Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hic1
Entrez Gene ID
100767977
Gene Name
hypermethylated in cancer 1
Gene Aliases
I79_014910
Chromosome Location
Chr.Un|NW_003614347: 791167 - 795903 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647845.1 XP_007646035.1 2 - 3 894 62

View Full Product Details