Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb16
Entrez Gene ID
100753821
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614071: 367353 - 374633 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504491.1 XP_003504539.1 3 - 4 1063 85

View Full Product Details