Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h6
Entrez Gene ID
100755342
Gene Name
zinc finger CCCH-type containing 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613665: 319799 - 351347 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007618341.1 XP_007616531.1 9 - 10 1358 75

View Full Product Details