Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfml1
Entrez Gene ID
100766460
Gene Name
olfactomedin-like 1
Gene Aliases
I79_021656
Chromosome Location
Chr.Un|NW_003615601: 48044 - 70455 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512807.1 XP_003512855.1 1 - 2 130 61

View Full Product Details