Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap2
Entrez Gene ID
100767422
Gene Name
A kinase (PRKA) anchor protein 2
Gene Aliases
I79_022621
Chromosome Location
Chr.Un|NW_003614309: 67491 - 101142 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647497.1 XP_007645687.1 1 - 2 2455 68

View Full Product Details