Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid3c
Entrez Gene ID
100753126
Gene Name
AT rich interactive domain 3C (BRIGHT-like)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614108: 1055464 - 1061250 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645484.1 XP_007643674.1 1 - 2 315 80

View Full Product Details