Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g2e
Entrez Gene ID
100753835
Gene Name
phospholipase A2, group IIE
Gene Aliases
I79_014439
Chromosome Location
Chr.Un|NW_003614302: 604904 - 609098 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506895.2 XP_003506943.1 3 - 4 392 100

View Full Product Details