Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1galt1
Entrez Gene ID
100761169
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
Gene Aliases
I79_010829
Chromosome Location
Chr.Un|NW_003614010: 1385933 - 1396547 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644439.1 XP_007642629.1 2 - 3 911 101
GenBank mRNA JP052144.1 - 1 - 1 913 101

View Full Product Details