Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc6a7
Entrez Gene ID
100764967
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614229: 426912 - 448177 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506321.2 XP_003506369.2 8 - 9 1170 64

View Full Product Details