Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc94
Entrez Gene ID
100755173
Gene Name
coiled-coil domain containing 94
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 161046 - 173329 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501742.2 XP_003501790.1 4 - 5 908 60
XM_007642404.1 XP_007640594.1 4 - 5 812 60
GenBank mRNA JI880583.1 - 3 - 4 351 60
JP041547.1 - 3 - 4 350 60

View Full Product Details