Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pus3
Entrez Gene ID
100759712
Gene Name
pseudouridylate synthase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613908: 1509005 - 1516353 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502705.2 XP_003502753.1 3 - 4 1031 150
XM_007643324.1 XP_007641514.1 3 - 4 1120 150
XM_007643325.1 XP_007641515.1 3 - 4 1184 150

View Full Product Details