Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
100758318
Gene Name
regulator of G-protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615486: 52194 - 89057 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653001.1 XP_007651191.1 3 - 4 236 81

View Full Product Details