Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hey1
Entrez Gene ID
100752534
Gene Name
hes-related family bHLH transcription factor with YRPW motif 1
Gene Aliases
I79_015033
Chromosome Location
Chr.Un|NW_003614364: 237459 - 313365 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647956.1 XP_007646146.1 3 - 4 439 91

View Full Product Details