Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kctd4
Entrez Gene ID
100768043
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613785: 276608 - 277387 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500297.2 XP_003500345.1 1 - 1 711 108

View Full Product Details