Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1D
Entrez Gene ID
102190816
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Gene Aliases
-
Chromosome Location
Chr.19: 11943631 - 12003680 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971835.2 XP_013827289.2 4 - 5 1246 64
XM_018064148.1 XP_017919637.1 4 - 5 1128 64

View Full Product Details